3-Chlor-2-fluorthioanisol

Lieferant: COMBI-BLOCKS

OT-0539-5G OT-0539-1G
COBBOT-0539-5GEA 937 EUR
COBBOT-0539-5G COBBOT-0539-1G
3-Chlor-2-fluorthioanisol
3-Chlor-2-fluorthioanisol
Formel: C₇H₆ClFS
Molecular Weight: 176.64 g/mol
MDL: MFCD09475846
CAS-Nummer: 214057-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR