N-Formyl-2-brom-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

OT-0547-1G
COBBOT-0547-1GEA 1400 EUR
COBBOT-0547-1G
N-Formyl-2-brom-4-(trifluormethoxy)anilin
N-Formyl-2-brom-4-(trifluormethoxy)anilin
Formel: C₈H₅BrF₃NO₂
MDL: MFCD09475870
CAS-Nummer: 941294-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR