4-Chlor-3-methylbenzotrifluorid

Lieferant: COMBI-BLOCKS

OT-0598-1G
COBBOT-0598-1GEA 153 EUR
COBBOT-0598-1G
4-Chlor-3-methylbenzotrifluorid
4-Chlor-3-methylbenzotrifluorid
Formel: C₈H₆ClF₃
Molecular Weight: 194,581 g/mol
MDL: MFCD09878417
CAS-Nummer: 97399-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR