2-Fluor-3-methylanisol

Lieferant: COMBI-BLOCKS

OT-0599-1G
COBBOT-0599-1GEA 0 EUR
COBBOT-0599-1G
2-Fluor-3-methylanisol
2-Fluor-3-methylanisol
Formel: C₈H₉FO
MDL: MFCD09878418
CAS-Nummer: 951885-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR