2-Chlor-5-(methylthio)benzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-methylthiobenzotrifluoride

OT-0616-1G
COBBOT-0616-1GEA 2850 EUR
COBBOT-0616-1G
2-Chlor-5-(methylthio)benzotrifluorid
2-Chlor-5-(methylthio)benzotrifluorid
Formel: C₈H₆ClF₃S
MDL: MFCD09878422
CAS-Nummer: 957066-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR