1-(3-Brompropoxy)-2-brombenzol

Lieferant: COMBI-BLOCKS

OT-0617-5G
COBBOT-0617-5GEA 353 EUR
COBBOT-0617-5G
1-(3-Brompropoxy)-2-brombenzol
1-(3-Brompropoxy)-2-brombenzol
Formel: C₉H₁₀Br₂O
MDL: MFCD02030868
CAS-Nummer: 37136-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR