2-Chlor-6-fluor-3-methoxyphenol

Lieferant: COMBI-BLOCKS

OT-0639-5G OT-0639-1G
COBBOT-0639-5GEA 0 EUR
COBBOT-0639-5G COBBOT-0639-1G
2-Chlor-6-fluor-3-methoxyphenol
2-Chlor-6-fluor-3-methoxyphenol
Formel: C₇H₆ClFO₂
Molecular Weight: 176.57 g/mol
MDL: MFCD09832266
CAS-Nummer: 1017777-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR