2-Brom-4-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-1-methoxy-4-nitrobenzene
OT-0648-25G
COBBOT-0648-25GEA
0
EUR
COBBOT-0648-25G
2-Brom-4-nitroanisol
2-Brom-4-nitroanisol
Formel:
C₇H₆BrNO₃ |
MDL:
MFCD00041242 CAS-Nummer: 5197-28-4 EINECS: 225-983-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...