1-(Methylsulfonyl)-4-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-(trifluoromethoxy)phenyl sulphone

OT-0649-5G OT-0649-25G OT-0649-1G
COBBOT-0649-5GEA 223 EUR
COBBOT-0649-5G COBBOT-0649-25G COBBOT-0649-1G
1-(Methylsulfonyl)-4-(trifluormethoxy)benzol
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Formel: C₈H₇F₃O₃S
MDL: MFCD00792426
CAS-Nummer: 87750-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR