1-(Methylsulfonyl)-4-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-(trifluoromethoxy)phenyl sulphone
OT-0649-5G
OT-0649-25G
OT-0649-1G
COBBOT-0649-5GEA
223
EUR
COBBOT-0649-5G
COBBOT-0649-25G
COBBOT-0649-1G
1-(Methylsulfonyl)-4-(trifluormethoxy)benzol
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Formel:
C₈H₇F₃O₃S |
MDL:
MFCD00792426 CAS-Nummer: 87750-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...