4-Brom-3-(trifluormethyl)anisol

Lieferant: COMBI-BLOCKS

OT-0668-25G OT-0668-1G OT-0668-5G
COBBOT-0668-25GEA 0 EUR
COBBOT-0668-25G COBBOT-0668-1G COBBOT-0668-5G
4-Brom-3-(trifluormethyl)anisol
4-Brom-3-(trifluormethyl)anisol
Formel: C₈H₆BrF₃O
MDL: MFCD08059527
CAS-Nummer: 400-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR