2-Brom-3-methylnaphthalin

Lieferant: COMBI-BLOCKS

OT-0691-100M
COBBOT-0691-100MEA 497 EUR
COBBOT-0691-100M
2-Brom-3-methylnaphthalin
2-Bromo-3-methylnaphthalene
Formel: C₁₁H₉Br
MDL: MFCD11504948
CAS-Nummer: 939-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR