2,4-Dichlor-5-methoxyphenol

Lieferant: COMBI-BLOCKS

OT-0695-1G
COBBOT-0695-1GEA 497 EUR
COBBOT-0695-1G
2,4-Dichlor-5-methoxyphenol
2,4-Dichlor-5-methoxyphenol
Formel: C₇H₆Cl₂O₂
MDL: MFCD10699699
CAS-Nummer: 18113-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR