2,6-Dibrom-4-nitrophenol

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Dibromo-2-hydroxy-5-nitrobenzene

OT-0697-25G OT-0697-5G OT-0697-100G
COBBOT-0697-25GEA 0 EUR
COBBOT-0697-25G COBBOT-0697-5G COBBOT-0697-100G
2,6-Dibrom-4-nitrophenol
2,6-Dibrom-4-nitrophenol
Formel: C₆H₃Br₂NO₃
Schmelzpunkt: 148,5 °C
MDL: MFCD00007334
CAS-Nummer: 99-28-5
EINECS: 202-744-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR