2,6-Dibrom-4-nitrophenol
Lieferant: COMBI-BLOCKS
Synonyme:
1,3-Dibromo-2-hydroxy-5-nitrobenzene
OT-0697-25G
OT-0697-5G
OT-0697-100G
COBBOT-0697-25GEA
0
EUR
COBBOT-0697-25G
COBBOT-0697-5G
COBBOT-0697-100G
2,6-Dibrom-4-nitrophenol
2,6-Dibrom-4-nitrophenol
Formel:
C₆H₃Br₂NO₃ Schmelzpunkt: 148,5 °C |
MDL:
MFCD00007334 CAS-Nummer: 99-28-5 EINECS: 202-744-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...