2-Brom-5-chlorphenol

Lieferant: COMBI-BLOCKS

OT-0707-1G
COBBOT-0707-1GEA 0 EUR
COBBOT-0707-1G
2-Brom-5-chlorphenol
2-Brom-5-chlorphenol
Formel: C₆H₄BrClO
MDL: MFCD00142870
CAS-Nummer: 13659-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR