2-(Trifluormethyl)anisol

Lieferant: COMBI-BLOCKS
Gefahr

OT-0710-5G OT-0710-1G
COBBOT-0710-5GEA 0 EUR
COBBOT-0710-5G COBBOT-0710-1G
2-(Trifluormethyl)anisol
2-(Trifluormethyl)anisol
Formel: C₈H₇F₃O
MDL: MFCD00142938
CAS-Nummer: 16222-42-7
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR