5-Brom-2-(trifluormethoxy)toluol

Lieferant: COMBI-BLOCKS

OT-0725-1G
COBBOT-0725-1GEA 175 EUR
COBBOT-0725-1G
5-Brom-2-(trifluormethoxy)toluol
5-Brom-2-(trifluormethoxy)toluol
Formel: C₈H₆BrF₃O
Molecular Weight: 255,04 g/mol
MDL: MFCD04972695
CAS-Nummer: 887268-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR