1-Boc-4-(3-bromphenoxy)piperidin

Lieferant: COMBI-BLOCKS

OT-0731-1G OT-0731-5G OT-0731-250M
COBBOT-0731-1GEA 486 EUR
COBBOT-0731-1G COBBOT-0731-5G COBBOT-0731-250M
1-Boc-4-(3-bromphenoxy)piperidin
1-Boc-4-(3-bromphenoxy)piperidin
Formel: C₁₆H₂₂BrNO₃
MDL: MFCD12026057
CAS-Nummer: 790667-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR