3-(4-Methoxyphenyl)propionsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(4-Methoxyphenyl)propansäure
OT-0749-5G
COBBOT-0749-5GEA
0
EUR
COBBOT-0749-5G
3-(4-Methoxyphenyl)propionsäure
3-(4-Methoxyphenyl)propionsäure
Formel:
C₁₀H₁₂O₃ Siedepunkt: 319 °C (1013 hPa) Schmelzpunkt: 95…102 °C |
MDL:
MFCD00002777 CAS-Nummer: 1929-29-9 EINECS: 217-678-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...