4-(4-Brom-1H-pyrazol-1-yl)-N-cyclohexylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0774-5G OT-0774-1G
COBBOT-0774-5GEA 0 EUR
COBBOT-0774-5G COBBOT-0774-1G
4-(4-Brom-1H-pyrazol-1-yl)-N-cyclohexylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-cyclohexylbenzolsulfonamid
Formel: C₁₅H₁₈BrN₃O₂S
MDL: MFCD12546462
CAS-Nummer: 1187385-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR