4-(4-Brom-1H-pyrazol-1-yl)-N-ethylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0781-1G OT-0781-5G
COBBOT-0781-1GEA 0 EUR
COBBOT-0781-1G COBBOT-0781-5G
4-(4-Brom-1H-pyrazol-1-yl)-N-ethylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-ethylbenzolsulfonamid
Formel: C₁₁H₁₂BrN₃O₂S
MDL: MFCD12756459
CAS-Nummer: 1187385-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR