1-Benzyloxy-4-fluorbenzol

Lieferant: COMBI-BLOCKS

OT-0795-5G OT-0795-25G OT-0795-100G
COBBOT-0795-5GEA 0 EUR
COBBOT-0795-5G COBBOT-0795-25G COBBOT-0795-100G
1-Benzyloxy-4-fluorbenzol
1-Benzyloxy-4-fluorbenzol
Formel: C₁₃H₁₁FO
MDL: MFCD03788273
CAS-Nummer: 370-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR