1-Benzyloxy-4-fluorbenzol
Lieferant: COMBI-BLOCKS
OT-0795-5G
OT-0795-25G
OT-0795-100G
COBBOT-0795-5GEA
0
EUR
COBBOT-0795-5G
COBBOT-0795-25G
COBBOT-0795-100G
1-Benzyloxy-4-fluorbenzol
1-Benzyloxy-4-fluorbenzol
Formel:
C₁₃H₁₁FO |
MDL:
MFCD03788273 CAS-Nummer: 370-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...