2,6-Dibrom-4-(trifluormethylsulfonyl)toluol

Lieferant: COMBI-BLOCKS

OT-0853-1G
COBBOT-0853-1GEA 404 EUR
COBBOT-0853-1G
2,6-Dibrom-4-(trifluormethylsulfonyl)toluol
2,6-Dibrom-4-(trifluormethylsulfonyl)toluol
Formel: C₈H₅Br₂F₃O₂S
MDL: MFCD12026060
CAS-Nummer: 1150271-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR