1-Brom-2-chlor-4-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

OT-0866-5G
COBBOT-0866-5GEA 0 EUR
COBBOT-0866-5G
1-Brom-2-chlor-4-(trifluormethoxy)benzol
Formel: C₇H₃BrClF₃O
MDL: MFCD11111892
CAS-Nummer: 892845-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR