2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid

Lieferant: COMBI-BLOCKS

OT-0898-25G OT-0898-5G
COBBOT-0898-25GEA 0 EUR
COBBOT-0898-25G COBBOT-0898-5G
2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid
2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid
Formel: C₇H₆Cl₂F₃NO
MDL: MFCD12546529
CAS-Nummer: 1187386-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR