1-((4-Fluorphenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

OT-0958-5G
COBBOT-0958-5GEA 149 EUR
COBBOT-0958-5G
1-((4-Fluorphenyl)sulfonyl)piperidin
1-((4-Fluorphenyl)sulfonyl)piperidin
Formel: C₁₁H₁₄FNO₂S
MDL: MFCD00194710
CAS-Nummer: 312-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR