4-Brom-2-fluorbenzamid

Lieferant: COMBI-BLOCKS

OT-1003-5G OT-1003-1G
COBBOT-1003-5GEA 449 EUR
COBBOT-1003-5G COBBOT-1003-1G
4-Brom-2-fluorbenzamid
4-Brom-2-fluorbenzamid
Formel: C₇H₅BrFNO
MDL: MFCD03094432
CAS-Nummer: 292621-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR