3-Brom-5-(trifluormethyl)phenol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-hydroxybenzotrifluoride

OT-1171-5G
COBBOT-1171-5GEA 0 EUR
COBBOT-1171-5G
3-Brom-5-(trifluormethyl)phenol
3-Brom-5-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
MDL: MFCD11041083
CAS-Nummer: 1025718-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR