4-Brom-2-(2-methoxyethoxy)anisol

Lieferant: COMBI-BLOCKS

OT-1173-5G
COBBOT-1173-5GEA 149 EUR
COBBOT-1173-5G
4-Brom-2-(2-methoxyethoxy)anisol
4-Brom-2-(2-methoxyethoxy)anisol
Formel: C₁₀H₁₃BrO₃
MDL: MFCD14584686
CAS-Nummer: 1132672-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR