4-Brom-3-iodtoluol

Lieferant: COMBI-BLOCKS

OT-1212-5G
COBBOT-1212-5GEA 0 EUR
COBBOT-1212-5G
4-Brom-3-iodtoluol
4-Brom-3-iodtoluol
Formel: C₇H₆BrI
MDL: MFCD07784420
CAS-Nummer: 858841-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR