1-Brom-2-chlor-5-fluor-4-nitrobenzol

Lieferant: COMBI-BLOCKS

OT-1250-1G
COBBOT-1250-1GEA 89.2 EUR
COBBOT-1250-1G
1-Brom-2-chlor-5-fluor-4-nitrobenzol
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene
Formel: C₆H₂BrClFNO₂
Molecular Weight: 254,4400 g/mol
MDL: MFCD11110551
CAS-Nummer: 1027833-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR