1-Boc-4-(4-brom-2-methoxyphenoxy)piperidin

Lieferant: COMBI-BLOCKS

OT-1254-1G
COBBOT-1254-1GEA 208 EUR
COBBOT-1254-1G
1-Boc-4-(4-brom-2-methoxyphenoxy)piperidin
1-Boc-4-(4-brom-2-methoxyphenoxy)piperidin
Formel: C₁₇H₂₄BrNO₄
MDL: MFCD16618960
CAS-Nummer: 1228957-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR