3-Brom-5-fluorbenzaldehyd

Lieferant: COMBI-BLOCKS

OT-1261-25G OT-1261-100G
COBBOT-1261-25GEA 301 EUR
COBBOT-1261-25G COBBOT-1261-100G
3-Brom-5-fluorbenzaldehyd
3-Bromo-5-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203.01 g/mol
MDL: MFCD04116319
CAS-Nummer: 188813-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR