2,6-Dibromtoluol

Lieferant: COMBI-BLOCKS
Achtung

OT-1437-100G
COBBOT-1437-100GEA 564 EUR
COBBOT-1437-100G
2,6-Dibromtoluol
2,6-Dibromtoluol
Formel: C₇H₆Br₂
Molecular Weight: 249.93 g/mol
MDL: MFCD00013524
CAS-Nummer: 69321-60-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR