1-(4-Bromphenyl)-2-pyrrolidinon

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Bromophenyl)pyrrolidin-2-one

OT-1465-1G
COBBOT-1465-1GEA 79.5 EUR
COBBOT-1465-1G
1-(4-Bromphenyl)-2-pyrrolidinon
1-(4-Bromphenyl)-2-pyrrolidinon
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,10 g/mol
MDL: MFCD00138502
CAS-Nummer: 7661-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR