4-Brom-1-(difluormethoxy)-2-fluorbenzol

Lieferant: COMBI-BLOCKS

OT-1620-1G
COBBOT-1620-1GEA 0 EUR
COBBOT-1620-1G
4-Brom-1-(difluormethoxy)-2-fluorbenzol
4-Brom-1-(difluormethoxy)-2-fluorbenzol
Formel: C₇H₄BrF₃O
MDL: MFCD11036067
CAS-Nummer: 147992-27-6
EINECS: 430-850-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR