2,5-Dichlor-2,5-dimethylhexan

Lieferant: COMBI-BLOCKS

OT-1806-100G
COBBOT-1806-100GEA 560 EUR
COBBOT-1806-100G
2,5-Dichlor-2,5-dimethylhexan
2,5-Dichloro-2,5-dimethylhexane
Formel: C₈H₁₆Cl₂
MDL: MFCD00126854
CAS-Nummer: 6223-78-5
EINECS: 228-310-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR