4-Brom-2-chlor-1-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
OT-1869-1G
COBBOT-1869-1GEA
0
EUR
COBBOT-1869-1G
4-Brom-2-chlor-1-(trifluormethoxy)benzol
4-Brom-2-chlor-1-(trifluormethoxy)benzol
Formel:
C₇H₃BrClF₃O |
MDL:
MFCD10699114 CAS-Nummer: 158579-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...