3-Brom-5-methoxybenzylalkohol

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-5-methoxyphenyl)methanol

OT-1946-1G
COBBOT-1946-1GEA 131 EUR
COBBOT-1946-1G
3-Brom-5-methoxybenzylalkohol
3-Brom-5-methoxybenzylalkohol
Formel: C₈H₉BrO₂
Molecular Weight: 217,599 g/mol
MDL: MFCD00229988
CAS-Nummer: 262450-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR