5-Brom-2,4-difluorbenzaldehyd

Lieferant: COMBI-BLOCKS

OT-2081-1G
COBBOT-2081-1GEA 51.3 EUR
COBBOT-2081-1G
5-Brom-2,4-difluorbenzaldehyd
5-Brom-2,4-difluorbenzaldehyd
Formel: C₇H₃BrF₂O
Molecular Weight: 220,9989 g/mol
MDL: MFCD09842439
CAS-Nummer: 473416-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR