2-Brom-3-methylanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-1-methoxy-3-methylbenzene

OT-2180-1G
COBBOT-2180-1GEA 102 EUR
COBBOT-2180-1G
2-Brom-3-methylanisol
2-Brom-3-methylanisol
Formel: C₈H₉BrO
Molecular Weight: 201,0600 g/mol
MDL: MFCD14705101
CAS-Nummer: 38197-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR