Chromon

Lieferant: COMBI-BLOCKS

Synonyme: 4H-Chromen-4-one

OT-2230-25G
COBBOT-2230-25GEA 246 EUR
COBBOT-2230-25G
Chromon
Chromon
Formel: C₉H₆O₂
Molecular Weight: 146,14 g/mol
MDL: MFCD00024064
CAS-Nummer: 491-38-3
EINECS: 207-737-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR