3-Phenylbenzoesäure

Lieferant: COMBI-BLOCKS

Synonyme: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

OT-8001-1G
COBBOT-8001-1GEA 25.9 EUR
COBBOT-8001-1G
3-Phenylbenzoesäure
3-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
MDL: MFCD00045846
CAS-Nummer: 716-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR