4-(2-Thienyl)anilin
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Thiophen-2-yl)aniline, 4-Thiophen-2-ylphenylamine
OT-8007-1G
COBBOT-8007-1GEA
191
EUR
COBBOT-8007-1G
4-(2-Thienyl)anilin
4-(2-Thienyl)anilin
Formel:
C₁₀H₉NS |
MDL:
MFCD01224064 CAS-Nummer: 70010-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...