4-(3,5-Bis(trifluormethyl)phenyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-3',5'-bis(trifluoromethyl)biphenyl

OT-8014-1G
COBBOT-8014-1GEA 21.6 EUR
COBBOT-8014-1G
4-(3,5-Bis(trifluormethyl)phenyl)anilin
4-(3,5-Bis(trifluormethyl)phenyl)anilin
Formel: C₁₄H₉F₆N
MDL: MFCD05154989
CAS-Nummer: 444143-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR