3-(3-Hydroxyphenyl)benzonitril
Lieferant: COMBI-BLOCKS
Synonyme:
3-Cyano-3'-hydroxybiphenyl
OT-8016-1G
COBBOT-8016-1GEA
154
EUR
COBBOT-8016-1G
3-(3-Hydroxyphenyl)benzonitril
3-(3-Hydroxyphenyl)benzonitril
Formel:
C₁₃H₉NO |
MDL:
MFCD05854256 CAS-Nummer: 154848-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...