4-Iod-o-anisidin

Lieferant: COMBI-BLOCKS

OT-8022-1G
COBBOT-8022-1GEA 0 EUR
COBBOT-8022-1G
4-Iod-o-anisidin
4-Iod-o-anisidin
Formel: C₇H₈INO
MDL: MFCD11044438
CAS-Nummer: 338454-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR