tert-Butyl-3-brombenzoat

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 3-Bromobenzoate

OT-8052-1G
COBBOT-8052-1GEA 38.5 EUR
COBBOT-8052-1G
tert-Butyl-3-brombenzoat
tert-Butyl-3-brombenzoat
Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257,1200 g/mol
MDL: MFCD06657802
CAS-Nummer: 69038-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR